Aldehyde PEG Aldehyde
Supplier: ALADDIN SCIENTIFIC
Synonyms: CHO-PEG-CHO
A163181-100MG A163181-500MG
ALA163181100MGEA 196.56 USD
ALA163181100MG ALA163181500MG
Aldehyde PEG Aldehyde
Aldehyde PEG Aldehyde
Heterobifunctional PEG derivative that can be used to modify proteins, peptides and other materials via amino or other acid reactive chemical groups. PEGylation can increase solubility and stability and reduce immunogenicity of peptides and proteins. It can also suppress the non-specific binding of charged molecules to the modified surfaces.