Acrylate PEG Biotin
Supplier: ALADDIN SCIENTIFIC
A163149-100MG A163149-1G A163149-500MG
ALA163149100MGEA 131.01 USD
ALA163149100MG ALA163149-1G ALA163149500MG
Acrylate PEG Biotin
Acrylate PEG Biotin
8 arm PEG derivatives have multiple reactive groups that can be used to modify proteins, peptides and other materials via their functional groups. PEGylation can increase solubility and stability and reduce immunogenicity of peptides and proteins. It can also suppress the non-specific binding of charged molecules to the modified surfaces. .