Acetylthio PEG amine
Supplier: ALADDIN SCIENTIFIC
A163145-100MG A163145-500MG
ALA163145100MGEA 132.91 USD
ALA163145100MG ALA163145500MG
Acetylthio PEG amine
Acetylthio PEG amine
8 arm PEG derivatives have multiple reactive groups that can be used to modify proteins, peptides and other materials via their functional groups. PEGylation can increase solubility and stability and reduce immunogenicity of peptides and proteins. It can also suppress the non-specific binding of charged molecules to the modified surfaces. .