8-Arm PEG Thiol
Supplier: ALADDIN SCIENTIFIC
Synonyms: 8 Arm PEG Thiol
A163133-100MG A163133-25MG
ALA163133100MGEA 184.21 USD
ALA163133100MG ALA163133-25MG
8-Arm PEG Thiol
8-Arm PEG Thiol
8 arm PEG derivatives have multiple reactive groups that can be used to modify proteins, peptides and other materials via their functional groups. PEGylation can increase solubility and stability and reduce immunogenicity of peptides and proteins. It can also suppress the non-specific binding of charged molecules to the modified surfaces. .