4-Arm PEG vinyl sulphone
Supplier: ALADDIN SCIENTIFIC
Synonyms:
4 Arm PEG vinyl sulphone
A163122-100MG
A163122-500MG
ALA163122100MGEA
262.11
USD
ALA163122100MG
ALA163122500MG
4-Arm PEG vinyl sulphone
4-Arm PEG vinyl sulphone
4 arm PEG derivatives have multiple reactive groups that can be used to modify proteins, peptides and other materials via their functional groups. PEGylation can increase solubility and stability and reduce immunogenicity of peptides and proteins. It can also suppress the non-specific binding of charged molecules to the modified surfaces. .