4-Arm PEG Biotin
Supplier: ALADDIN SCIENTIFIC
Synonyms: 4 Arm PEG Biotin, 4arm-PEG-NH2, 4arm-PEG10K-NH2
A163119-1G A163119-250MG
ALA163119-1GEA 552.81 USD
ALA163119-1G ALA163119250MG
4-Arm PEG Biotin
4-Arm PEG Biotin
4 arm PEG derivatives have multiple reactive groups that can be used to modify proteins, peptides and other materials via their functional groups. PEGylation can increase solubility and stability and reduce immunogenicity of peptides and proteins. It can also suppress the non-specific binding of charged molecules to the modified surfaces. .