2-Arm/Branched PEG Biotin
Supplier: ALADDIN SCIENTIFIC
Synonyms:
2 Arm/Branched PEG Biotin
A163090-100MG
A163090-500MG
ALA163090100MGEA
131.01
USD
ALA163090100MG
ALA163090500MG
2-Arm/Branched PEG Biotin
2-Arm/Branched PEG Biotin
2 arm PEG derivatives have two PEG branches with one or two functional groups that can be used to modify proteins, peptides and other materials via their functional groups. PEGylation can increase solubility and stability and reduce immunogenicity of peptides and proteins. It can also suppress the non-specific binding of charged molecules to the modified surfaces. .