Amantadine sulfate ≥98%
Supplier: ALADDIN SCIENTIFIC
Synonyms:
1-Adamantanammonium sulfate, 1-Adamantanammonium sulphate, 1-Aminoadamantane sulfate, 1-Adamantanamine sulphate, 1-Adamantylamine sulfate, 1-Adamantylamine sulphate, 1-Adamantanamine sulfate, 1-Aminoadamantane sulphate
A151773-25G
A151773-5G
ALA151773-25GEA
25.56
USD
ALA151773-25G
ALA151773-5G
Amantadine sulfate ≥98%
Amantadine sulphate
Formula:
C₁₀H₁₉NO₄S MW: 386.58 g/mol |
MDL Number:
MFCD00077251 CAS Number: 31377-23-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...