ɑ-(Trichloromethyl)benzyl acetate ≥98% (by GC)

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 2,2,2-Trichloro-1-phenylethyl acetate, Acetic acid 2,2,2-Trichloro-1-phenylethyl Ester, Acetic Acid ɑ-(Trichloromethyl)benzyl Ester, Acetic Acid 2,2,2-Trichloro-1-phenylethyl Ester, 2,2,2-Trichloro-1-phenylethyl Acetate, Acetic acid α-(Trichloromethyl)benzyl Ester, α-(Trichloromethyl)benzyl Acetate

A151755-250G A151755-25G
ALA151755-250GEA 67.36 USD
ALA151755-250G ALA151755-25G
ɑ-(Trichloromethyl)benzyl acetate ≥98% (by GC)
alpha-(Trichloromethyl)benzyl acetate
Formula: C₁₀H₉Cl₃O₂
Boiling Pt: 282°C
Melting Pt: 88 °C
MDL Number: MFCD00000804
CAS Number: 90-17-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3e1c2faa17ae6a7e347a3276c9351a1c350949bc&vid=3e1c2faa17ae6a7e347a3276c9351a1c350949bc&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR