4'-Acetoxyacetophenone ≥98% (by GC)

Supplier: ALADDIN SCIENTIFIC

Synonyms: 4-Acetylphenyl acetate, 4-Acetoxyacetophenone, p-Acetylphenyl acetate

A151754-100G A151754-25G A151754-5G
ALA151754-100GEA 74.96 USD
ALA151754-100G ALA151754-25G ALA151754-5G
4'-Acetoxyacetophenone ≥98% (by GC)
4'-Acetoxyacetophenone
Formula: C₁₀H₁₀O₃
MW: 178.19 g/mol
Boiling Pt: 165-170°C
Melting Pt: 52-56°C
MDL Number: MFCD00017229
CAS Number: 13031-43-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=496a0c9b4cacfdec89d6a7e13fe0be63b096ee25&vid=496a0c9b4cacfdec89d6a7e13fe0be63b096ee25&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR