5-Amino-2-bromo-3-methylpyridine ≥98% (by GC)
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
5-Amino-2-bromo-3-picoline
A151752-1G
A151752-250MG
A151752-5G
ALA151752-1GEA
23.66
USD
ALA151752-1G
ALA151752250MG
ALA151752-5G
5-Amino-2-bromo-3-methylpyridine ≥98% (by GC)
5-Amino-2-bromo-3-methylpyridine
Formula:
C₆H₇BrN₂ MW: 187.04 g/mol Melting Pt: 100 °C |
MDL Number:
MFCD03095085 CAS Number: 38186-83-3 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...