4'-Acetamidoacetophenone ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 4-Acetamidoacetophenone, N-(4-Acetylphenyl)acetamide

A151745-1G A151745-25G A151745-50G A151745-5G
ALA151745-1GEA 9.41 USD
ALA151745-1G ALA151745-25G ALA151745-50G ALA151745-5G
4'-Acetamidoacetophenone ≥98%
4'-Acetamidoacetophenone
Formula: C₁₀H₁₁NO₂
Melting Pt: 166-172°C
Storage Temperature: Ambient
MDL Number: MFCD00014965
CAS Number: 2719-21-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6fd1858ac23c0fbd94368272299596400e401054&vid=6fd1858ac23c0fbd94368272299596400e401054&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR