5-Acetyl-2-chloropyridine ≥98%

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: 3-Acetyl-6-chloropyridine

A151743-1G A151743-5G
ALA151743-1GEA 35.06 USD
ALA151743-1G ALA151743-5G
5-Acetyl-2-chloropyridine ≥98%
5-Acetyl-2-chloropyridine
Formula: C₇H₆ClNO
MW: 155.5816 g/mol
Melting Pt: 102 °C
MDL Number: MFCD03407343
CAS Number: 55676-22-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2927ad0b23c4721699af34482eb506873230a1f4&vid=2927ad0b23c4721699af34482eb506873230a1f4&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR