Acetylenedicarboxylic acid monopotassium salt ≥95% (by titration)

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: Potassium Hydrogen Acetylenedicarboxylate, Acetylenedicarboxylic Acid Monopotassium Salt, Monopotassium Acetylenedicarboxylate, mono-Potassium 2-butynedioate

A151734-100G A151734-25G A151734-500G A151734-5G
ALA151734-100GEA 64.51 USD
ALA151734-100G ALA151734-25G ALA151734-500G ALA151734-5G
Acetylenedicarboxylic acid monopotassium salt ≥95% (by titration)
Acetylenedicarboxylic acid monopotassium salt
Formula: C₄H₂O₄
MW: 152.15 g/mol
MDL Number: MFCD00013143
CAS Number: 928-04-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1e08b3e2b0e0232100030f2ca23028f2dd1ed9b0&vid=1e08b3e2b0e0232100030f2ca23028f2dd1ed9b0&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR