4-Amino-1-isopropylpiperidine ≥98%
Supplier: ALADDIN SCIENTIFIC
|
Danger
|
A151715-1G
A151715-25G
A151715-5G
ALA151715-1GEA
23.66
USD
ALA151715-1G
ALA151715-25G
ALA151715-5G
4-Amino-1-isopropylpiperidine ≥98%
4-Amino-1-isopropylpiperidine
Formula:
C₈H₁₈N₂ Density: 0.903 g/cm³ Flash Pt: 59 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03411606 CAS Number: 127285-08-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...