Acid violet 43 ≥97% (by HPLC)

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: Acid Violet 43

A151709-1G A151709-5G
ALA151709-1GEA 74.01 USD
ALA151709-1G ALA151709-5G
Acid violet 43 ≥97% (by HPLC)
Acid violet 43
Formula: C₂₁H₁₅NO₆S
MDL Number: MFCD00068446
CAS Number: 4430-18-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=54003a4af4305758c9aac2a818b5aba6b24e6a21&vid=54003a4af4305758c9aac2a818b5aba6b24e6a21&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR