3-Aminocoumarin ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 3-Amino-2H-chromen-2-one

A151702-100MG A151702-1G A151702-250MG
ALA151702100MGEA 29.36 USD
ALA151702100MG ALA151702-1G ALA151702250MG
3-Aminocoumarin ≥98%
3-Aminocoumarin
Formula: C₉H₇NO₂
Melting Pt: 137 °C
MDL Number: MFCD00016965
CAS Number: 1635-31-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d0fefc8d8093dc9e96ddeebfb220d893b70e3fcb&vid=d0fefc8d8093dc9e96ddeebfb220d893b70e3fcb&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR