7-Acetoxy-4-methylcoumarin ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 4-Methyl-2-oxo-2H-chromen-7-yl acetate, 4-Methylumbelliferyl acetate

A151699-100G A151699-25G A151699-5G
ALA151699-100GEA 614.56 USD
ALA151699-100G ALA151699-25G ALA151699-5G
7-Acetoxy-4-methylcoumarin ≥98%
7-Acetoxy-4-methylcoumarin
Formula: C₁₂H₁₀O₄
MW: 218.21 g/mol
Melting Pt: 147-152° C
Storage Temperature: Freezer
MDL Number: MFCD00006865
CAS Number: 2747-05-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR