4-Amino-o-anisic acid ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 4-Amino-2-methoxybenzenecarboxylic acid

A151694-100G A151694-25G A151694-5G
ALA151694-100GEA 201.31 USD
ALA151694-100G ALA151694-25G ALA151694-5G
4-Amino-o-anisic acid ≥98%
4-Amino-o-anisic acid
Formula: C₈H₉NO₃
MW: 167.16
Melting Pt: 145 °C
Storage Temperature: Ambient
MDL Number: MFCD00114479
CAS Number: 2486-80-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a4aa64e5c482ede59a67731fc3501df9a3c7d80d&vid=a4aa64e5c482ede59a67731fc3501df9a3c7d80d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR