4-Pentyloxybenzaldehyde ≥98% (by GC)

Supplier: ALADDIN SCIENTIFIC

Synonyms: 4-(Pentyloxy)benzaldehyde, 4-n-Pentyloxybenzaldehyde, 4-Amyloxybenzaldehyde, 4-n-Amyloxybenzaldehyde

A151683-25G A151683-5G
ALA151683-25GEA 46.46 USD
ALA151683-25G ALA151683-5G
4-Pentyloxybenzaldehyde ≥98% (by GC)
4-Pentyloxybenzaldehyde
Formula: C₁₂H₁₆O₂
Boiling Pt: 158°C
Density: 1.02 g/cm³
Flash Pt: 129 °C
MDL Number: MFCD00014135
CAS Number: 5736-91-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2c2a6ac5154beea3adc9e097316f639cf651a999&vid=2c2a6ac5154beea3adc9e097316f639cf651a999&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR