5,6,7,7a-Tetrahydrothieno[3,2-c]pyridin-2(4H)-one hydrochloride ≥97% (by titration)
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
5,6,7,7a-Tetrahydrothieno[3,2-c]pyridin-2(4H)-one Hydrochloride
A151681-1G
A151681-25G
A151681-5G
ALA151681-1GEA
79.71
USD
ALA151681-1G
ALA151681-25G
ALA151681-5G
5,6,7,7a-Tetrahydrothieno[3,2-c]pyridin-2(4H)-one hydrochloride ≥97% (by titration)
5,6,7,7a-Tetrahydrothieno[3,2-c]pyridin-2(4H)-one hydrochloride
Formula:
C₇H₁₀ClNOS Melting Pt: 210 °C Storage Temperature: Freezer |
MDL Number:
MFCD11111130 CAS Number: 115473-15-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...