4-(3-Aminophenyl)-2-methyl-3-butyn-2-ol ≥98% (by HPLC)

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 3-(3-Hydroxy-3-methyl-1-butyn-1-yl)aniline

A151680-1G A151680-25G A151680-5G
ALA151680-1GEA 22.71 USD
ALA151680-1G ALA151680-25G ALA151680-5G
4-(3-Aminophenyl)-2-methyl-3-butyn-2-ol ≥98% (by HPLC)
4-(3-Aminophenyl)-2-methyl-3-butyn-2-ol
Formula: C₁₁H₁₃NO
MW: 175.23 g/mol
Melting Pt: 118-122°C
MDL Number: MFCD06200785
CAS Number: 69088-96-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d494b3a6b3e2e0a25f8f13f2833a741fd4afcdae&vid=d494b3a6b3e2e0a25f8f13f2833a741fd4afcdae&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR