2-Acetamido-5-bromopyridine ≥98% (by GC)

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 2-Acetylamino-5-bromopyridine, N-(5-Bromopyridin-2-yl)acetamide

A151676-10G A151676-1G A151676-5G
ALA151676-10GEA 44.56 USD
ALA151676-10G ALA151676-1G ALA151676-5G
2-Acetamido-5-bromopyridine ≥98% (by GC)
2-Acetamido-5-bromopyridine
Formula: C₇H₇BrN₂O
Melting Pt: 174-179°C
Storage Temperature: Ambient
MDL Number: MFCD00468968
CAS Number: 7169-97-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c685987eef0c55f3807a4c30f3bfe423db3f5afc&vid=c685987eef0c55f3807a4c30f3bfe423db3f5afc&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR