2-Amino-3,5-dichloro-4-methylpyridine ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 3,5-Dichloro-4-methylpyridin-2-amine

A151673-1G A151673-200MG
ALA151673-1GEA 18.91 USD
ALA151673-1G ALA151673200MG
2-Amino-3,5-dichloro-4-methylpyridine ≥97%
2-Amino-3,5-dichloro-4-methylpyridine
Formula: C₆H₆Cl₂N₂
MW: 177.0300 g/mol
Melting Pt: 123-127°C
MDL Number: MFCD00129031
CAS Number: 31430-47-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=98ec159548b0f7055ea42bf4fcfac75747322723&vid=98ec159548b0f7055ea42bf4fcfac75747322723&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR