3-Amino-2-pyridone ≥96% (by HPLC)

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: 3-Amino-2-hydroxypyridine, 3-Amino-2-pyridinol, 3-Amino-2-pyridinone

A151666-1G A151666-25G A151666-5G
ALA151666-1GEA 70.21 USD
ALA151666-1G ALA151666-25G ALA151666-5G
3-Amino-2-pyridone ≥96% (by HPLC)
3-Amino-2-pyridone
Formula: C₅H₆N₂O
Melting Pt: 124 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03411556
CAS Number: 33630-99-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=91680f5a3a64049a37b08515e843aede9070c753&vid=91680f5a3a64049a37b08515e843aede9070c753&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR