5-Amino-2-methoxypyridine ≥98% (by GC)
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
A151664-100G
A151664-25G
A151664-5G
ALA151664-100GEA
234.56
USD
ALA151664-100G
ALA151664-25G
ALA151664-5G
5-Amino-2-methoxypyridine ≥98% (by GC)
5-Amino-2-methoxypyridine
Product Application:5-Amino-2-methoxypyridine is used as a reagent in the synthesis of a novel series of thienopyrimidines as highly potent and selective PI3K inhibitors. 5-Amino-2-methoxypyridine is also used to synthesize EMPA; a novel high-affinity selective antagonist for the OX2 receptor.
Formula:
C₆H₈N₂O MW: 124.14 Boiling Pt: 265 Melting Pt: 29-31°C Flash Pt: >110°C Storage Temperature: Refrigerator |
MDL Number:
MFCD00006264 CAS Number: 6628-77-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...