6-Aminonicotinonitrile ≥98% (by GC)

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: 6-Amino-3-pyridinecarbonitrile, 6-aminopyridine-3-carbonitrile, 2-Amino-5-cyanopyridine, 2-Aminopyridine-5-carbonitrile

A151661-100G A151661-1G A151661-25G A151661-5G
ALA151661-100GEA 432.16 USD
ALA151661-100G ALA151661-1G ALA151661-25G ALA151661-5G
6-Aminonicotinonitrile ≥98% (by GC)
6-Aminonicotinonitrile
Formula: C₆H₅N₃
MW: 119.13 g/mol
Boiling Pt: 250 °C
Melting Pt: 164
MDL Number: MFCD00128902
CAS Number: 4214-73-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3df05c4f82f416941b57bcb1e92fa7832014dd8e&vid=3df05c4f82f416941b57bcb1e92fa7832014dd8e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR