(1ɑ,3ɑ,5ɑ)-1,3,5-Cyclohexanetricarbonitrile ≥98%

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: (1α,3α,5α)-Cyclohexane-1,3,5-tricarbonitrile, (1alpha,3alpha,5alpha)-1,3,5-Tricyanocyclohexane, ['(1α,3α,5α)-1,3,5-Tricyanocyclohexane'], (1α,3α,5α)-1,3,5-Cyclohexanetricarbonitrile

A151647-1G A151647-250MG
ALA151647-1GEA 261.16 USD
ALA151647-1G ALA151647250MG
(1ɑ,3ɑ,5ɑ)-1,3,5-Cyclohexanetricarbonitrile ≥98%
(1alpha,3alpha,5alpha)-1,3,5-Cyclohexanetricarbonitrile
Formula: C₉H₉N₃
MW: 159.19 g/mol
Melting Pt: 170 °C
MDL Number: MFCD08276327
CAS Number: 168280-46-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fedb7b6024d6d4def1b0c2b41f74d56a4607ab24&vid=fedb7b6024d6d4def1b0c2b41f74d56a4607ab24&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR