5-Amino-2-(hydroxymethyl)phenylboronic acid cyclic monoester ≥95% (by HPLC)
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
6-amino-1-hydroxy-2,1-benzoxaborolane, 5-amino-2-hydroxymethyl-phenylboronic acid, 6-aminobenzo[c]-[1,2]oxaborol-1(3H)-ol, 1-hydroxy-3H-2,1-benzoxaborol-6-amine, 5-Amino-2-(hydroxymethyl)phenylboronic Acid Cyclic Monoester, 6-amino-3H-benzo[c][1,2]oxaborol-1-ol, 6-Amino-1,3-dihydro-1-hydroxy-2,1-benzoxaborole, 6-Aminobenzo[c][1,2]oxaborol-1(3H)-ol, 6-amino-2,1-benzoxaborol-1(3H)-ol
A151641-1G
A151641-250MG
ALA151641-1GEA
404.61
USD
ALA151641-1G
ALA151641250MG
5-Amino-2-(hydroxymethyl)phenylboronic acid cyclic monoester ≥95% (by HPLC)
5-Amino-2-(hydroxymethyl)phenylboronic acid cyclic monoester
Formula:
C₇H₈BNO₂ MW: 148.96 g/mol Melting Pt: 148°C Storage Temperature: Freezer |
MDL Number:
MFCD14584713 CAS Number: 117098-94-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...