3-Aminobenzylamine ≥98% (by GC)

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: alpha-Amino-m-toluidine, 3-(Aminomethyl)aniline

A151635-100G A151635-25G A151635-5G
ALA151635-100GEA 399.86 USD
ALA151635-100G ALA151635-25G ALA151635-5G
3-Aminobenzylamine ≥98% (by GC)
3-Aminobenzylamine
Formula: C₇H₁₀N₂
MW: 122.17 g/mol
Boiling Pt: 134°C
Melting Pt: 41-45°C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00078355
CAS Number: 4403-70-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4cec63da00777d7eebc4f75e6933e97008672ce2&vid=4cec63da00777d7eebc4f75e6933e97008672ce2&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR