2-Acetoxyisobutyroyl chloride ≥97% (by titration)

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: α-Acetoxyisobutyryl chloride,2-Acetoxy-2-methylpropionyl chloride, 1-Chloro-2-methyl-1-oxopropan-2-yl acetate, 2-Acetoxyisobutyryl Chloride, Chlorocarbonyl-1-methylethyl acetate

A151622-25G A151622-5G
ALA151622-25GEA 74.01 USD
ALA151622-25G ALA151622-5G
2-Acetoxyisobutyroyl chloride ≥97% (by titration)
2-Acetoxyisobutyroyl chloride
Formula: C₆H₉ClO₃
Boiling Pt: 70°C
Density: 1.15 g/cm³
Flash Pt: 68°C
MDL Number: MFCD00000708
CAS Number: 40635-66-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c264fd0e62afbf1bdf3861a370403d609e23bc62&vid=c264fd0e62afbf1bdf3861a370403d609e23bc62&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR