Azelaoyl chloride ≥98% (by GC, titration analysis)

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: Azelaoyl Chloride, Azelaoylchloride

A151621-25ML A151621-5ML
ALA151621-25MLEA 91.11 USD
ALA151621-25ML ALA151621-5ML
Azelaoyl chloride ≥98% (by GC, titration analysis)
Azelaoyl chloride
Formula: C₉H₁₄Cl₂O₂
MW: 225.11 g/mol
Boiling Pt: 172°C
Flash Pt: >113 °C
Storage Temperature: Ambient
MDL Number: MFCD00000767
CAS Number: 123-98-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e7fe0acf45c3f694473a8eb49793b04ecb2a3b33&vid=e7fe0acf45c3f694473a8eb49793b04ecb2a3b33&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR