2-Aminoisonicotinamide ≥98% (by HPLC)

Supplier: ALADDIN SCIENTIFIC
Danger

A151619-1G A151619-25G A151619-5G
ALA151619-1GEA 37.91 USD
ALA151619-1G ALA151619-25G ALA151619-5G
2-Aminoisonicotinamide ≥98% (by HPLC)
2-Aminoisonicotinamide

Application:2-Aminopyridine-4-carboxamide is used as pharmaceutical intermediate.

Formula: C₆H₇N₃O
Melting Pt: 252-258°C
MDL Number: MFCD03791262
CAS Number: 13538-42-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=88521eefbe422f7c34d375dc8cbbcb6d87680848&vid=88521eefbe422f7c34d375dc8cbbcb6d87680848&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR