2-Aminophenol hydrochloride ≥98% (by HPLC)
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
2-Aminophenol Hydrochloride
A151616-1G
A151616-5G
ALA151616-1GEA
10.36
USD
ALA151616-1G
ALA151616-5G
2-Aminophenol hydrochloride ≥98% (by HPLC)
2-Aminophenol hydrochloride
Formula:
C₆H₇NO MW: 145.59 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00035471 CAS Number: 51-19-4 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...