2-Aminophenol-4-sulfonamide ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 3-Amino-4-hydroxybenzenesulphonamide, 3-Amino-4-hydroxybenzenesulfonamide

A151612-100G A151612-25G
ALA151612-100GEA 49.31 USD
ALA151612-100G ALA151612-25G
2-Aminophenol-4-sulfonamide ≥97%
2-Aminophenol-4-sulphonamide
Formula: C₆H₈N₂O₃S
Melting Pt: 199 °C
Storage Temperature: Ambient
MDL Number: MFCD00035782
CAS Number: 98-32-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8b1167429ed05bad310f025fb8336eb91ce0927a&vid=8b1167429ed05bad310f025fb8336eb91ce0927a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR