2-Amino-4,5-dimethylbenzoic acid ≥95%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 4,5-Dimethylanthranilic acid, 2-Amino-4,5-dimethylbenzoic Acid

A151593-1G A151593-200MG A151593-25G
ALA151593-1GEA 309.61 USD
ALA151593-1G ALA151593200MG ALA151593-25G
2-Amino-4,5-dimethylbenzoic acid ≥95%
2-Amino-4,5-dimethylbenzoic acid
Formula: C₉H₁₁NO₂
Melting Pt: 208 °C
MDL Number: MFCD00156998
CAS Number: 15089-51-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6bf0bc60574673b2f0c990ca8c8a6b198268d930&vid=6bf0bc60574673b2f0c990ca8c8a6b198268d930&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR