3-Amino-2-bromopyridine ≥98% (by GC, titration analysis)

Supplier: ALADDIN SCIENTIFIC
Danger

A151581-100G A151581-1G A151581-25G A151581-5G
ALA151581-100GEA 2131.71 USD
ALA151581-100G ALA151581-1G ALA151581-25G ALA151581-5G
3-Amino-2-bromopyridine ≥98% (by GC, titration analysis)
3-Amino-2-bromopyridine

Employed in a palladium-catalyzed synthesis of indole systems containing fused medium- and large-ring heterocycles.

Formula: C₅H₅BrN₂
Melting Pt: 79°C
MDL Number: MFCD00234064
CAS Number: 39856-58-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9c5173e56bd69e8590f9284e2c73a1930b659277&vid=9c5173e56bd69e8590f9284e2c73a1930b659277&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR