2-Amino-3,5-dichloropyridine ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 3,5-Dichloro-2-pyridylamine, 3,5-Dichloropyridin-2-amine

A151577-100G A151577-250G A151577-25G A151577-5G
ALA151577-100GEA 264.01 USD
ALA151577-100G ALA151577-250G ALA151577-25G ALA151577-5G
2-Amino-3,5-dichloropyridine ≥98%
2-Amino-3,5-dichloropyridine

2-Amino-3,5-dichloropyridine was used in the synthesis of bis(2-amino-3,5-dihalopyridinium) tetrahalocuprate(II) compounds and 2-fluoro-3,5-dichloropyridine.

Formula: C₅H₄Cl₂N₂
Melting Pt: 81-83°C
MDL Number: MFCD00006313
CAS Number: 4214-74-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=91ab97f548e67863113471d919fd25c9efedf9fa&vid=91ab97f548e67863113471d919fd25c9efedf9fa&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR