2,6-Dihydroxy-9,10-anthraquinone ≥95%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 2,6-Dihydroxyanthracene-9,10-dione, Anthraflavic acid, 2,6-Dihydroxyanthraquinone

A151573-1G A151573-5G
ALA151573-1GEA 56.91 USD
ALA151573-1G ALA151573-5G
2,6-Dihydroxy-9,10-anthraquinone ≥95%
2,6-Dihydroxy-9,10-anthraquinone
Formula: C₁₄H₈O₄
Melting Pt: 330°C
MDL Number: MFCD00001228
CAS Number: 84-60-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2449e57004513a218c08d37dbbb44ce62fe5bd16&vid=2449e57004513a218c08d37dbbb44ce62fe5bd16&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR