4-Amino-3-bromobenzonitrile ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A151560-1G A151560-250MG A151560-25G A151560-5G
ALA151560-1GEA 11.31 USD
ALA151560-1G ALA151560250MG ALA151560-25G ALA151560-5G
4-Amino-3-bromobenzonitrile ≥98%
4-Amino-3-bromobenzonitrile
Formula: C₇H₅BrN₂
Melting Pt: 106-110°C
MDL Number: MFCD01861386
CAS Number: 50397-74-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=69b155d5fc87588876a8bec9b2e4b661fb99dc4a&vid=69b155d5fc87588876a8bec9b2e4b661fb99dc4a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR