ɑ,ɑ'-Bis(4-hydroxyphenyl)-1,4-diisopropylbenzene ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A151532-100G A151532-25G A151532-5G
ALA151532-100GEA 146.21 USD
ALA151532-100G ALA151532-25G ALA151532-5G
ɑ,ɑ'-Bis(4-hydroxyphenyl)-1,4-diisopropylbenzene ≥98%
alpha,alpha'-Bis(4-hydroxyphenyl)-1,4-diisopropylbenzene
Formula: C₂₄H₂₆O₂
Melting Pt: 195 °C
MDL Number: MFCD00191315
CAS Number: 2167-51-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=08d2701e0c2140e9ded0af9bb481ffb91a9ba551&vid=08d2701e0c2140e9ded0af9bb481ffb91a9ba551&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR