2-Aminoethanethiol-p-toluenesulfonate ≥98%

Supplier: ALADDIN SCIENTIFIC

Synonyms: 2-Aminoethanethiol p-Toluenesulfonate, 2-Aminoethanethiol p-Toluenesulphonate, 2-Mercaptoethylamine p-Toluenesulfonate, Cysteamine p-Toluenesulfonate, Cysteamine p-Toluenesulphonate, 2-Mercaptoethylamine p-Toluenesulphonate

A151526-5G A151526-1G
ALA151526-5GEA 125.31 USD
ALA151526-5G ALA151526-1G
2-Aminoethanethiol-p-toluenesulfonate ≥98%
2-Aminoethanethiol-p-toluenesulfonate
Formula: C₂H₇NS
Melting Pt: 166.0 to 170.0 °C
Storage Temperature: Ambient
MDL Number: MFCD00060252
CAS Number: 3037-04-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR