2-Amino-9,9-dimethylfluorene ≥99%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 9,9-Dimethyl-9H-fluoren-2-amine

A151524-100G A151524-1G A151524-250MG A151524-25G
ALA151524-100GEA 3172.91 USD
ALA151524-100G ALA151524-1G ALA151524250MG ALA151524-25G
2-Amino-9,9-dimethylfluorene ≥99%
2-Amino-9,9-dimethylfluorene
Formula: C₁₅H₁₅N
Melting Pt: 166.0 to 170.0 °C
Density: 1.107 g/cm³
MDL Number: MFCD09953790
CAS Number: 108714-73-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c21c458983535052dca0cc3a0625d51a9ef22680&vid=c21c458983535052dca0cc3a0625d51a9ef22680&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR