1-Amino-4-bromoanthraquinone-2-sulfonic acid sodium salt ≥98% (by HPLC)
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
1-Amino-4-bromo-9,10-dihydro-9,10-dioxo-2-anthracenesulfonic acid sodium salt, 1-Amino-4-bromoanthraquinone-2-sulphonic Acid Sodium Salt, Sodium 1-amino-4-bromo-9,10-dioxo-9,10-dihydroanthracene-2-sulfonate, Bromaminic acid Sodium Salt, Bromaminic acid sodium salt, 1-Amino-4-bromoanthraquinone-2-sulfonic Acid Sodium Salt, Sodium 1-amino-4-bromo-9,10-dioxo-9,10-dihydroanthracene-2-sulphonate
A151521-100G
A151521-25G
A151521-5G
ALA151521-100GEA
140.51
USD
ALA151521-100G
ALA151521-25G
ALA151521-5G
1-Amino-4-bromoanthraquinone-2-sulfonic acid sodium salt ≥98% (by HPLC)
1-Amino-4-bromoanthraquinone-2-sulfonic acid sodium salt
Formula:
C₁₄H₈BrNO₅S Melting Pt: >300 °C |
MDL Number:
MFCD00019160 CAS Number: 6258-06-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...