1-Allyl-3,5-dimethylpyrazole ≥97% (by GC)

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 1-Allyl-3,5-dimethyl-1H-pyrazole

A151512-25G A151512-5G A151512-1G
ALA151512-25GEA 418.86 USD
ALA151512-25G ALA151512-5G ALA151512-1G
1-Allyl-3,5-dimethylpyrazole ≥97% (by GC)
1-Allyl-3,5-dimethylpyrazole
Formula: C₈H₁₂N₂
Boiling Pt: 192°C
Density: 0.94 g/cm³
Flash Pt: 80°C
MDL Number: MFCD00034407
CAS Number: 13369-74-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=35ed1adf0ca369fa625dbdcca7c964581b36dc57&vid=35ed1adf0ca369fa625dbdcca7c964581b36dc57&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR