3-Amino-4-bromo-5-methylpyrazole ≥98% (by HPLC, titration analysis)

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 4-Bromo-5-methyl-1H-pyrazol-3-amine

A151508-1G A151508-250MG A151508-5G
ALA151508-1GEA 55.96 USD
ALA151508-1G ALA151508250MG ALA151508-5G
3-Amino-4-bromo-5-methylpyrazole ≥98% (by HPLC, titration analysis)
3-Amino-4-bromo-5-methylpyrazole
Formula: C₄H₆BrN₃
MW: 176.02 g/mol
Melting Pt: 120 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03412169
CAS Number: 1780-72-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b50a14c84caedd4f2ca1b4f1b1dc76fa01f79ba6&vid=b50a14c84caedd4f2ca1b4f1b1dc76fa01f79ba6&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR