4-Azido-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-homoalanine ≥98% (by HPLC)
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
A151486-250MG
A151486-50MG
ALA151486250MGEA
197.51
USD
ALA151486250MG
ALA151486-50MG
4-Azido-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-homoalanine ≥98% (by HPLC)
4-Azido-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-homoalanine
Formula:
C₁₉H₁₈N₄O₄ Melting Pt: 127 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD11052920 CAS Number: 942518-20-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...