5-Amino-2-methoxybenzoic acid ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 5-Amino-2-methoxybenzoic Acid

A151475-1G A151475-250MG A151475-5G
ALA151475-1GEA 37.91 USD
ALA151475-1G ALA151475250MG ALA151475-5G
5-Amino-2-methoxybenzoic acid ≥97%
5-Amino-2-methoxybenzoic acid
Formula: C₈H₉NO₃
Melting Pt: 161-166 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09260887
CAS Number: 3403-47-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=264c3ed0859932b99fac194ed80a614402d044d7&vid=264c3ed0859932b99fac194ed80a614402d044d7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR