2-Amino-4,4,4-trifluorobutyric acid ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: (±)-2-Amino-4,4,4-trifluorobutyric acid, 2-Amino-4,4,4-trifluorobutanoic acid

A151473-100MG A151473-1G A151473-250MG
ALA151473100MGEA 28.41 USD
ALA151473100MG ALA151473-1G ALA151473250MG
2-Amino-4,4,4-trifluorobutyric acid ≥98%
(±)-2-Amino-4,4,4-trifluorobutyric acid
Formula: C₄H₆F₃NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD00041417
CAS Number: 15959-93-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1102f01ab4534c612d5f1abdc59f0742a34d2e6c&vid=1102f01ab4534c612d5f1abdc59f0742a34d2e6c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR