5-Amino-2,4,6-triiodoisophthalic acid ≥96%

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: 5-Amino-2,4,6-triiodoisophthalic Acid

A151465-100G A151465-25G A151465-50G A151465-5G
ALA151465-100GEA 240.26 USD
ALA151465-100G ALA151465-25G ALA151465-50G ALA151465-5G
5-Amino-2,4,6-triiodoisophthalic acid ≥96%
5-Amino-2,4,6-triiodoisophthalic acid
Formula: C₈H₄I₃NO₄
Melting Pt: 270 °C
MDL Number: MFCD00190167
CAS Number: 35453-19-1
EINECS: 252-575-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c38e2784de5fb96d61eb94392836285b76527858&vid=c38e2784de5fb96d61eb94392836285b76527858&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR